Assay Details
Target Gene Details
Entrez Gene ID: | 16907 |
Gene Name: | lamin B2 |
Gene Aliases: |
- |
Location: |
Chr.10:80901363-80918245 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmnb2 | NM_010722.5 | 12 | 3119 | NP_034852.2 |
| AK012232.1 | 1 | 717 | ||
| BC042430.1 | 12 | 3124 | AAH42430.1 | |
| BC051985.1 | 12 | 3095 | AAH51985.1 | |
| X54098.1 | 12 | 3066 | CAA38032.1 |

Genomic Map