Assay Details
Target Gene Details
Entrez Gene ID: | 215890 |
Gene Name: | clavesin 2 |
Gene Aliases: |
A330019N05Rik, Rlbp1l2 |
Location: |
Chr.10:33512286-33624905 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clvs2 | NM_175448.3 | 2 | 725 | NP_780657.1 |
| AK039308.1 | 2 | 726 | BAC30312.1 | |
| AK049896.1 | 2 | 286 | BAC33976.1 | |
| AK049988.1 | 2 | 286 | ||
| AK053797.1 | 2 | 748 | BAC35529.1 | |
| AK139409.1 | 2 | 800 | BAE23998.1 | |
| AK139868.1 | 2 | 778 | ||
| AK163214.1 | 2 | 746 | ||
| AK169009.1 | 2 | 895 | BAE40806.1 | |
| BC058539.1 | 2 | 732 | AAH58539.1 |
More Information
Set Membership: |
|

Genomic Map