Assay Details
Target Gene Details
Entrez Gene ID: | 26968 |
Gene Name: | immunoglobulin superfamily containing leucine-rich repeat |
Gene Aliases: |
AW553146 |
Location: |
Chr.9:58156246-58159239 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Islr | NM_001195431.1 | 2 | 1762 | NP_001182360.1 |
| NM_012043.4 | 2 | 1894 | NP_036173.1 | |
| AB024538.1 | 2 | 1715 | BAA85972.1 | |
| AI838480.1 | ||||
| BC006602.1 | 2 | 1815 | AAH06602.1 |

Genomic Map