Assay Details
Target Gene Details
Entrez Gene ID: | 235627 |
Gene Name: | neurobeachin-like 2 |
Gene Aliases: |
1110014F23Rik, BC042396, mKIAA0540 |
Location: |
Chr.9:110624789-110654161 on Build GRCm38 |
Assay Gene Location: | Within Exon 54 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbeal2 | NM_183276.2 | 54 | 8434 | NP_899099.2 |
| AK003694.2 | 5 | 610 | BAC25048.1 | |
| AK129158.1 | 29 | 4702 | BAC97968.2 | |
| BC157956.1 | 54 | 8415 | ||
| BC172118.1 | 53 | 8334 |

Genomic Map