Assay Details
Target Gene Details
Entrez Gene ID: | 19823 |
Gene Name: | ring finger protein 7 |
Gene Aliases: |
Rbx2, SAG |
Location: |
Chr.9:96470957-96478701 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf7 | NM_011279.2 | 3 | 386 | NP_035409.1 |
| AF092877.1 | 3 | 389 | AAD25961.1 | |
| AK002628.1 | 3 | 385 | ||
| AK003248.1 | 3 | 387 | BAB22666.1 | |
| AK003963.1 | 3 | 386 | ||
| AK006703.1 | 3 | 466 | ||
| AK007588.1 | 3 | 372 | ||
| AK011328.1 | 3 | 385 | ||
| AK146030.1 | 3 | 387 | BAE26843.1 | |
| AK209943.1 | ||||
| BC011127.1 | 3 | 384 | AAH11127.1 |
More Information
Set Membership: |
|

Genomic Map