Assay Details
Target Gene Details
Entrez Gene ID: | 12566 |
Gene Name: | cyclin-dependent kinase 2 |
Gene Aliases: |
A630093N05Rik |
Location: |
Chr.10:128697938-128705051 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk2 | NM_016756.4 | 1 | 279 | NP_058036.1 |
| NM_183417.3 | 1 | 279 | NP_904326.1 | |
| AJ223732.1 | 1 | 181 | CAA11533.1 | |
| AK036589.1 | 1 | 232 | ||
| AK042462.1 | 1 | 232 | ||
| AK146845.1 | 1 | 259 | BAE27476.1 | |
| AK148017.1 | 1 | 218 | BAE28290.1 | |
| AK151979.1 | 1 | 229 | BAE30846.1 | |
| AK152922.1 | 1 | 232 | BAE31597.1 | |
| AK153983.1 | 1 | 231 | BAE32294.1 | |
| AK154997.1 | 1 | 231 | BAE32983.1 | |
| AK187629.1 | ||||
| AK189612.1 | ||||
| BC005654.1 | 1 | 240 | AAH05654.1 | |
| CN705407.1 | 1 | 279 | ||
| U63337.1 | 1 | 207 | AAB37128.1 |
More Information
Set Membership: |
|

Genomic Map