Assay Details
Target Gene Details
Entrez Gene ID: | 56484 |
Gene Name: | forkhead box O3 |
Gene Aliases: |
1110048B16Rik, 2010203A17Rik, C76856, FKHRL1, Fkhr2, Foxo3a |
Location: |
Chr.10:42181845-42276746 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxo3 | NM_019740.2 | 2 | 331 | NP_062714.1 |
| AF114259.1 | 2 | 331 | AAD42107.1 | |
| AK047413.1 | 2 | 289 | BAC33049.1 | |
| AK139629.1 | 2 | 317 | ||
| AK143198.1 | 2 | 349 | ||
| BC019532.1 | 1 | 171 |

Genomic Map