Assay Details
Target Gene Details
Entrez Gene ID: | 78801 |
Gene Name: | adenylate kinase 7 |
Gene Aliases: |
4930502N02Rik, AK 7 |
Location: |
Chr.12:105703859-105782447 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak7 | NM_030187.1 | 18 | 2835 | NP_084463.1 |
| AK019664.1 | 18 | 2700 | BAB31828.1 | |
| AK029555.1 | 18 | 2839 |
More Information
Set Membership: |
|

Genomic Map