Assay Details
Target Gene Details
Entrez Gene ID: | 72729 |
Gene Name: | CDC42 small effector 2 |
Gene Aliases: |
2810404F18Rik, AA408783, AA536669, SPEC2 |
Location: |
Chr.11:54717415-54787703 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42se2 | NM_178626.3 | 6 | 2751 | NP_848741.1 |
| AK088090.1 | 6 | 2667 | BAC40139.1 | |
| AK090381.1 | 6 | 2686 | BAC41193.1 | |
| AK135775.1 | 6 | 2702 | BAE22655.1 | |
| AK145931.1 | 6 | 2717 | BAE26762.1 | |
| AK146313.1 | 6 | 2666 | BAE27068.1 | |
| AK160934.1 | 6 | 2736 | BAE36099.1 | |
| BC048935.1 | 6 | 2622 | AAH48935.1 | |
| BY688282.1 |

Genomic Map