Assay Details
Target Gene Details
Entrez Gene ID: | 13543 |
Gene Name: | dishevelled 2, dsh homolog (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.11:70000626-70010109 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dvl2 | NM_007888.3 | 15 | 2777 | NP_031914.3 |
| AK144001.1 | 15 | 2808 | ||
| AK146822.1 | 15 | 2777 | BAE27460.1 | |
| AK159895.1 | 15 | 2610 | BAE35461.1 | |
| AK168376.1 | 15 | 2685 | BAE40307.1 | |
| AK197190.1 | 1 | 42 | ||
| BC053050.1 | 15 | 2691 | AAH53050.1 | |
| U24160.1 | 15 | 2789 | AAC52827.1 |
More Information
Set Membership: |
|

Genomic Map