Assay Details
Target Gene Details
Entrez Gene ID: | 103551 |
Gene Name: | RIKEN cDNA E130012A19 gene |
Gene Aliases: |
AA409164, AI413509, AW539173, E13, esPRC2p48 |
Location: |
Chr.11:97627387-97629716 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E130012A19Rik | NM_175332.3 | 1 | 1165 | NP_780541.2 |
| AK087411.1 | 1 | 1152 | BAC39864.1 | |
| BC006054.1 | 1 | 604 | AAH06054.1 | |
| BC055770.1 | 1 | 1165 | AAH55770.1 |

Genomic Map