Assay Details
Target Gene Details
Entrez Gene ID: | 14428 |
Gene Name: | galanin receptor 2 |
Gene Aliases: |
mGalR |
Location: |
Chr.11:116280232-116287498 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galr2 | NM_010254.4 | 2 | 1767 | NP_034384.3 |
| AK053776.1 | 2 | 1768 | BAC35518.1 | |
| AK132050.1 | 2 | 1575 | ||
| AK137830.1 | 1 | 596 | ||
| AW046646.1 | 1 | 191 |
More Information
Set Membership: |
|

Genomic Map