Assay Details
Target Gene Details
Entrez Gene ID: | 78926 |
Gene Name: | growth arrest-specific 2 like 1 |
Gene Aliases: |
4930500E24Rik, D0Jmb1, Gar22, TU-71.1 |
Location: |
Chr.11:5054131-5065327 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas2l1 | NM_001190406.1 | NP_001177335.1 | ||
| NM_001190408.1 | NP_001177337.1 | |||
| NM_030228.3 | NP_084504.2 | |||
| NM_144560.3 | NP_653146.1 | |||
| AF508323.1 | 4 | 1198 | AAM34262.1 | |
| AF508324.1 | AAM34263.1 | |||
| AK019661.1 | BAB31826.1 | |||
| AK052229.1 | ||||
| AK082864.1 | ||||
| BC031785.1 | AAH31785.1 |
More Information
Set Membership: |
|

Genomic Map