Assay Details
Target Gene Details
Entrez Gene ID: | 237891 |
Gene Name: | growth arrest-specific 2 like 2 |
Gene Aliases: |
- |
Location: |
Chr.11:83421635-83429521 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas2l2 | NM_001013759.2 | 6 | 2430 | NP_001013781.1 |
| BC147480.1 | 6 | 2430 | ||
| BC150847.1 | 6 | 2430 |

Genomic Map