Assay Details
Target Gene Details
Entrez Gene ID: | 192786 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 6 |
Gene Aliases: |
A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052 |
Location: |
Chr.11:54521754-54699286 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgef6 | NM_001252494.1 | 20 | 2958 | NP_001239423.1 |
| NM_001252496.1 | 20 | 2958 | NP_001239425.1 | |
| NM_175258.4 | 19 | 2943 | NP_780467.2 | |
| AK143331.1 | 7 | 1330 | BAE25343.1 | |
| AK220459.1 | 8 | 1346 | BAD90488.1 | |
| BC025553.1 | 2 | 437 | AAH25553.1 | |
| BC092525.1 | 2 | 510 | AAH92525.1 | |
| BC141182.1 | 20 | 2857 |
More Information
Set Membership: |
|

Genomic Map