Assay Details
Target Gene Details
Entrez Gene ID: | 268417 |
Gene Name: | zinc finger with KRAB and SCAN domains 17 |
Gene Aliases: |
Nizp1, Zfp496, Znf496 |
Location: |
Chr.11:59485520-59506640 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zkscan17 | NM_001130529.2 | 10 | 3633 | NP_001124001.1 |
| NM_001291014.1 | 10 | 3387 | NP_001277943.1 | |
| NM_172941.4 | 10 | 3680 | NP_766529.3 | |
| AK051711.1 | 10 | 2969 | BAC34730.1 | |
| AK155196.1 | 10 | 3210 | BAE33110.1 | |
| AK155490.1 | 10 | 3262 | BAE33290.1 | |
| AK157002.1 | 4 | 1342 | BAE33928.1 | |
| AK170924.1 | 10 | 3210 | BAE42117.1 | |
| AK170989.1 | 10 | 3215 | BAE42163.1 | |
| AK171190.1 | 10 | 3262 | BAE42300.1 | |
| AK219529.1 | 1 | 332 | ||
| AY302242.1 | 9 | 3161 | AAQ73562.1 | |
| BC040205.1 | 9 | 3165 | AAH40205.1 | |
| CR519620.1 |
More Information
Set Membership: |
|

Genomic Map