Assay Details
Target Gene Details
Entrez Gene ID: | 67178 |
Gene Name: | zinc finger, matrin type 5 |
Gene Aliases: |
2610510L01Rik, D11Bwg0572e, D11Bwg1548e |
Location: |
Chr.11:4704606-4737669 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmat5 | NM_026015.2 | 6 | 611 | NP_080291.1 |
| AK003361.1 | 6 | 604 | BAB22736.1 | |
| AK007565.1 | 6 | 598 | BAB25111.1 | |
| AK012098.1 | 6 | 612 | BAB28029.1 | |
| AK013415.1 | 5 | 584 | BAB28843.1 | |
| BC031796.1 | 6 | 590 | AAH31796.1 |

Genomic Map