Assay Details
Target Gene Details
Entrez Gene ID: | 217473 |
Gene Name: | ankyrin repeat and MYND domain containing 2 |
Gene Aliases: |
AI035571, ZMYND20 |
Location: |
Chr.12:36157124-36197291 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankmy2 | NM_146033.3 | 10 | 2269 | NP_666145.3 |
| AK077556.1 | 10 | 2240 | BAC36860.1 | |
| AK164434.1 | 10 | 2171 | BAE37787.1 | |
| BF453918.1 | 1 | 190 |

Genomic Map