Assay Details
Target Gene Details
Entrez Gene ID: | 11799 |
Gene Name: | baculoviral IAP repeat-containing 5 |
Gene Aliases: |
AAC-11, Api4, TIAP, survivin40 |
Location: |
Chr.11:117849237-117855743 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Birc5 | NM_001012273.1 | 3 | 2897 | NP_001012273.1 |
| NM_009689.2 | NP_033819.1 | |||
| AB013819.1 | BAA28266.1 | |||
| AF077349.1 | AAD34225.1 | |||
| AK038229.1 | 1 | 997 | ||
| AK142768.1 | BAE25189.1 | |||
| BC004702.1 | 3 | 2815 | AAH04702.1 | |
| CN724527.1 | ||||
| EU366955.1 |

Genomic Map