Assay Details
Target Gene Details
Entrez Gene ID: | 207165 |
Gene Name: | bromodomain PHD finger transcription factor |
Gene Aliases: |
9430093H17Rik, Falz |
Location: |
Chr.11:107033081-107131922 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bptf | NM_176850.2 | 6 | 2210 | NP_789820.2 |
| BB325728.2 | ||||
| BC037661.1 | 7 | 2044 | ||
| CF981870.1 | 3 | 445 | ||
| CN724595.1 | ||||
| GU189417.1 | 6 | 1763 |

Genomic Map