Assay Details
Target Gene Details
Entrez Gene ID: | 12189 |
Gene Name: | breast cancer 1 |
Gene Aliases: |
- |
Location: |
Chr.11:101488754-101551956 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brca1 | NM_009764.3 | 10 | 3632 | NP_033894.3 |
| AB221610.1 | 1 | 861 | ||
| BC068303.1 | 10 | 3468 | AAH68303.1 | |
| U31625.1 | 10 | 3489 | AAB17114.1 | |
| U32446.1 | 10 | 3542 | AAA96393.1 | |
| U35641.1 | 10 | 3525 | AAB17113.1 | |
| U36475.1 | 9 | 3452 | AAC52323.1 | |
| U68174.1 | 10 | 3536 | AAD00168.1 |
More Information
Set Membership: |
|

Genomic Map