Assay Details
Target Gene Details
Entrez Gene ID: | 71962 |
Gene Name: | GATS protein-like 3 |
Gene Aliases: |
2410008K03Rik |
Location: |
Chr.11:4218162-4222409 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gatsl3 | NM_028022.1 | 9 | 1261 | NP_082298.1 |
| AK010455.1 | 9 | 1244 | BAB26953.1 | |
| BC039988.1 | 9 | 1261 | AAH39988.1 |

Genomic Map