Assay Details
Target Gene Details
Entrez Gene ID: | 103988 |
Gene Name: | glucokinase |
Gene Aliases: |
GLK, Gk, Gls006, HK4, HKIV, HXKP, Hlb62, MODY2 |
Location: |
Chr.11:5900816-5950081 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gck | NM_001287386.1 | 10 | 1544 | NP_001274315.1 |
| NM_010292.5 | 10 | 1902 | NP_034422.2 | |
| BC011139.1 | AAH11139.1 | |||
| L38990.1 | 10 | 1420 | AAB00360.1 |
More Information
Set Membership: |
|

Genomic Map