Assay Details
Target Gene Details
Entrez Gene ID: | 50932 |
Gene Name: | misshapen-like kinase 1 (zebrafish) |
Gene Aliases: |
B55, MINK, Map4k6, Ysk2 |
Location: |
Chr.11:70562594-70614482 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mink1 | NM_001045959.1 | 16 | 2159 | NP_001039424.1 |
| NM_001045964.1 | 16 | 2159 | NP_001039429.1 | |
| NM_016713.2 | 16 | 2159 | NP_057922.2 | |
| NM_176893.2 | 16 | 2159 | NP_795712.2 | |
| AB035697.1 | 16 | 2144 | BAA90752.1 | |
| AB041925.1 | 16 | 2145 | BAA94837.1 | |
| BC011346.1 | 11 | 1531 | AAH11346.1 | |
| BC052474.1 | 16 | 2014 | AAH52474.1 | |
| BU707794.1 | ||||
| CF736764.1 |
More Information
Set Membership: |
|

Genomic Map