Assay Details
Target Gene Details
Entrez Gene ID: | 67139 |
Gene Name: | MIS12 homolog (yeast) |
Gene Aliases: |
2510025F08Rik, AU044651, Misc12 |
Location: |
Chr.11:71019611-71027371 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mis12 | NM_025993.2 | 3 | 2085 | NP_080269.1 |
| AK010995.1 | 3 | 2086 | BAB27314.1 | |
| AK077702.1 | 1 | 962 | ||
| AK132010.1 | 3 | 2087 | BAE20937.1 | |
| AK154531.1 | 3 | 2086 | BAE32656.1 | |
| BC026790.1 | 3 | 1771 | AAH26790.1 |

Genomic Map