Assay Details
Target Gene Details
Entrez Gene ID: | 18016 |
Gene Name: | neurofibromatosis 2 |
Gene Aliases: |
merlin |
Location: |
Chr.11:4757650-4849544 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf2 | NM_001252250.1 | 1 | 353 | NP_001239179.1 |
| NM_001252251.1 | 1 | 353 | NP_001239180.1 | |
| NM_001252252.1 | 1 | 353 | NP_001239181.1 | |
| NM_010898.4 | 1 | 353 | NP_035028.2 | |
| AK045998.1 | 1 | 175 | BAC32567.1 | |
| AK197717.1 | ||||
| BC005442.1 | 1 | 219 | AAH05442.1 | |
| BY144572.1 | ||||
| BY271441.1 | 1 | 313 | ||
| L28176.1 | 1 | 125 | AAA39808.1 | |
| X74671.1 | 1 | 359 | CAA52737.1 |
More Information
Set Membership: |
|

Genomic Map