Assay Details
Target Gene Details
Entrez Gene ID: | 18016 |
Gene Name: | neurofibromatosis 2 |
Gene Aliases: |
merlin |
Location: |
Chr.11:4757650-4849544 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf2 | NM_001252250.1 | 17 | 2753 | NP_001239179.1 |
| NM_001252252.1 | 16 | 2724 | NP_001239181.1 | |
| NM_010898.4 | 16 | 2708 | NP_035028.2 | |
| AK161134.1 | 1 | 3327 | ||
| AK167685.1 | 13 | 1799 | BAE39732.1 | |
| BC005442.1 | AAH05442.1 | |||
| L27090.1 | 16 | 2159 | AAA63648.1 | |
| L27105.1 | 17 | 2204 | AAA39807.1 | |
| X75759.1 | 6 | 997 | CAA53386.1 |
More Information
Set Membership: |
|

Genomic Map