Assay Details
Target Gene Details
Entrez Gene ID: | 18472 |
Gene Name: | platelet-activating factor acetylhydrolase, isoform 1b, subunit 1 |
Gene Aliases: |
LIS-1, Lis1, MMS10-U, Mdsh, Ms10u, Pafaha |
Location: |
Chr.11:74673949-74724384 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pafah1b1 | NM_013625.4 | 11 | 2360 | NP_038653.1 |
| NR_037610.2 | 11 | 2633 | ||
| AK147207.1 | 11 | 2298 | BAE27764.1 | |
| AK147229.1 | 9 | 5413 | ||
| AK147237.1 | 11 | 2352 | BAE27787.1 | |
| AK147309.1 | 10 | 4010 | BAE27837.1 | |
| AK147507.1 | 11 | 2634 | BAE27961.1 | |
| AK147515.1 | 11 | 2347 | BAE27965.1 | |
| AK147533.1 | 11 | 2327 | BAE27977.1 | |
| AK147793.1 | 11 | 2022 | BAE28141.1 | |
| AK152284.1 | 1 | 401 | ||
| AK161751.1 | ||||
| AK164276.1 | 1 | 478 | ||
| AK168929.1 | 11 | 2000 | BAE40742.1 | |
| L25108.1 | 11 | 2260 | ||
| U95116.1 | 11 | 2655 | AAC04610.1 |

Genomic Map