Assay Details
Target Gene Details
Entrez Gene ID: | 22658 |
Gene Name: | polycomb group ring finger 2 |
Gene Aliases: |
Mel18, Rnf110, Zfp144, mel-18 |
Location: |
Chr.11:97688821-97701344 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcgf2 | NM_001163307.1 | 11 | 1724 | NP_001156779.1 |
| NM_001163308.1 | 10 | 1586 | NP_001156780.1 | |
| NM_009545.2 | 13 | 1991 | NP_033571.1 | |
| AK142837.1 | 10 | 1499 |

Genomic Map