Assay Details
Target Gene Details
Entrez Gene ID: | 19401 |
Gene Name: | retinoic acid receptor, alpha |
Gene Aliases: |
Nr1b1, RAR, RARalpha1 |
Location: |
Chr.11:98927809-98974942 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rara | NM_001177302.1 | 3 | 410 | NP_001170773.1 |
| NM_001177303.1 | 2 | 155 | NP_001170774.1 | |
| NM_009024.2 | 2 | 277 | NP_033050.2 | |
| AK042163.1 | 2 | 156 | BAE20623.1 | |
| AK154623.1 | 2 | 250 | BAE32722.1 | |
| BC010216.1 | 2 | 275 | AAH10216.1 | |
| S56656.1 | 2 | 257 | AAB25783.1 | |
| X56572.1 | 2 | 298 | CAA39919.1 |
More Information
Set Membership: |
|

Genomic Map