Assay Details
Target Gene Details
Entrez Gene ID: | 19401 |
Gene Name: | retinoic acid receptor, alpha |
Gene Aliases: |
Nr1b1, RAR, RARalpha1 |
Location: |
Chr.11:98927809-98974942 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rara | NM_001176528.1 | 8 | 2163 | NP_001169999.1 |
| NM_001177302.1 | 10 | 2368 | NP_001170773.1 | |
| NM_001177303.1 | 9 | 2113 | NP_001170774.1 | |
| NM_009024.2 | 9 | 2235 | NP_033050.2 | |
| AK042163.1 | 9 | 2114 | BAE20623.1 | |
| AK153644.1 | 8 | 1709 | BAE32132.1 | |
| AK154623.1 | 9 | 2208 | BAE32722.1 | |
| BC010216.1 | 9 | 2233 | AAH10216.1 |
More Information
Set Membership: |
|

Genomic Map