Assay Details
Target Gene Details
Entrez Gene ID: | 19416 |
Gene Name: | RAS, dexamethasone-induced 1 |
Gene Aliases: |
Dexras1 |
Location: |
Chr.11:59963181-59965080 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasd1 | NM_009026.4 | 1 | 211 | NP_033052.1 |
| AF009246.1 | 1 | 215 | AAC53538.1 | |
| AK038932.1 | 1 | 214 | BAC30173.1 | |
| BC034166.1 | 1 | 185 | AAH34166.1 | |
| CT010376.1 | 1 | 74 | CAJ18583.1 |

Genomic Map