Assay Details
Target Gene Details
Entrez Gene ID: | 78926 |
Gene Name: | growth arrest-specific 2 like 1 |
Gene Aliases: |
4930500E24Rik, D0Jmb1, Gar22, TU-71.1 |
Location: |
Chr.11:5054131-5065327 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas2l1 | NM_001190406.1 | NP_001177335.1 | ||
| NM_030228.3 | NP_084504.2 | |||
| AK019661.1 | BAB31826.1 | |||
| AK082864.1 |
Target Gene Details
Entrez Gene ID: | 75668 |
Gene Name: | RAS-like, family 10, member A |
Gene Aliases: |
2210403B10Rik, AI852688 |
Location: |
Chr.11:5057981-5060383 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasl10a | NM_145216.3 | 3 | 1009 | NP_660251.1 |
| AK008807.1 | 3 | 1010 | ||
| AK044011.1 | 3 | 1010 | BAC31738.1 | |
| AV078172.1 | 1 | 130 | ||
| BC107200.1 | AAI07201.1 | |||
| BC107201.1 | AAI07202.1 |
More Information
Set Membership: |
|

Genomic Map