Assay Details
Target Gene Details
Entrez Gene ID: | 22368 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 2 |
Gene Aliases: |
GRC, OTRPC2, VRL-1, Vrl1 |
Location: |
Chr.11:62574486-62600305 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv2 | NM_011706.2 | 3 | 412 | NP_035836.2 |
| AB021665.1 | 3 | 496 | BAA78478.1 | |
| AK089004.1 | 4 | 427 | BAC40695.1 | |
| AK134052.1 | 2 | 725 | ||
| AK162841.1 | 2 | 286 | ||
| AK171222.1 | 2 | 335 | ||
| AK200065.1 | ||||
| AK209862.1 | ||||
| BB631765.1 | 3 | 412 | ||
| BC005415.1 | 2 | 359 | AAH05415.1 |
More Information
Set Membership: |
|

Genomic Map