Assay Details
Target Gene Details
Entrez Gene ID: | 66152 |
Gene Name: | ubiquinol-cytochrome c reductase, complex III subunit X |
Gene Aliases: |
1110020P15Rik, AA960494, Ucrc |
Location: |
Chr.11:4701968-4704362 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcr10 | NM_197979.2 | 2 | 320 | NP_932096.1 |
| AK002227.1 | 2 | 310 | ||
| AK003881.1 | 2 | 317 | ||
| AK012489.1 | 2 | 313 | ||
| AK168491.1 | 2 | 294 | BAE40378.1 | |
| AW985484.1 | 2 | 288 | ||
| BC024518.1 | 2 | 304 | AAH24518.1 | |
| BC087874.1 | 2 | 310 | AAH87874.1 | |
| BC100591.1 | 2 | 310 | AAI00592.1 | |
| BX522336.1 | 3 | 370 | ||
| CB599515.1 | 2 | 323 |
More Information
Set Membership: |
|

Genomic Map