Assay Details
Target Gene Details
Entrez Gene ID: | 11541 |
Gene Name: | adenosine A2b receptor |
Gene Aliases: |
A2BAR, A2BR, A2b, AA2BR, AI480866, AI605384 |
Location: |
Chr.11:62248984-62266452 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adora2b | NM_007413.4 | 2 | 914 | NP_031439.2 |
| AK047002.1 | 2 | 915 | BAC32938.1 | |
| AK077289.1 | 2 | 688 | BAC36732.1 | |
| AK080759.1 | 2 | 909 | ||
| BC116415.1 | 2 | 872 | ||
| BC116416.1 | 2 | 872 | ||
| U05673.1 | 2 | 829 | AAA19001.1 |
More Information
Set Membership: |
|

Genomic Map