Assay Details
Target Gene Details
Entrez Gene ID: | 11548 |
Gene Name: | adrenergic receptor, alpha 1b |
Gene Aliases: |
[a]1b |
Location: |
Chr.11:43774605-43901251 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra1b | NM_001284380.1 | 3 | 370 | NP_001271309.1 |
| NM_001284381.1 | 4 | 520 | NP_001271310.1 | |
| NM_007416.4 | 1 | 366 | NP_031442.2 | |
| AK004891.1 | 4 | 477 | BAB23647.1 | |
| AK043877.1 | 1 | 211 | BAC31691.1 | |
| AK139448.1 | 1 | 367 | ||
| BC018330.1 | 3 | 343 | ||
| BC037002.1 | 3 | 344 | AAH37002.1 | |
| BC138653.1 | 1 | 84 | ||
| BC138654.1 | 1 | 84 | ||
| BY107791.1 | ||||
| Y12738.1 | 1 | 690 | CAA73272.1 |
More Information
Set Membership: |
|

Genomic Map