Assay Details
Target Gene Details
Entrez Gene ID: | 11548 |
Gene Name: | adrenergic receptor, alpha 1b |
Gene Aliases: |
[a]1b |
Location: |
Chr.11:43774605-43901251 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra1b | NM_001284380.1 | 4 | 2906 | NP_001271309.1 |
| NM_001284381.1 | 5 | 3056 | NP_001271310.1 | |
| NM_007416.4 | 2 | 2902 | NP_031442.2 | |
| AK043877.1 | 2 | 2747 | BAC31691.1 | |
| CA319459.1 | 1 | 274 |
More Information
Set Membership: |
|

Genomic Map