Assay Details
Target Gene Details
Entrez Gene ID: | 15114 |
Gene Name: | huntingtin-associated protein 1 |
Gene Aliases: |
HAP-1 |
Location: |
Chr.11:100347327-100356353 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hap1 | NM_010404.3 | 1 | 303 | NP_034534.1 |
| NM_177981.2 | 1 | 303 | NP_817090.1 | |
| AJ000262.1 | 1 | 249 | CAA03978.1 | |
| AJ002271.1 | 1 | 249 | CAA05286.1 | |
| BC034089.1 | 1 | 241 | AAH34089.1 | |
| BC053043.1 | 1 | 290 | AAH53043.1 | |
| BY288360.1 | 1 | 304 |
More Information
Set Membership: |
|

Genomic Map