Assay Details
Target Gene Details
Entrez Gene ID: | 268482 |
Gene Name: | keratin 12 |
Gene Aliases: |
AI835216, K12, Krt-12, Krt1-12 |
Location: |
Chr.11:99415664-99422593 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt12 | NM_010661.2 | 1 | 485 | NP_034791.2 |
| AK053362.1 | 1 | 486 | ||
| AK143182.1 | 1 | 482 | ||
| BC131911.1 | 1 | 481 | ||
| CK627700.1 | 1 | 471 | ||
| U02880.1 | 1 | 482 | AAA17792.1 |

Genomic Map