Assay Details
Target Gene Details
Entrez Gene ID: | 16669 |
Gene Name: | keratin 19 |
Gene Aliases: |
AI663979, EndoC, K19, Krt-1.19, Krt1-19 |
Location: |
Chr.11:100140810-100145919 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt19 | NM_008471.2 | 6 | 1199 | NP_032497.1 |
| AB472075.1 | 6 | 1199 | ||
| BC034561.1 | 6 | 1192 | AAH34561.1 | |
| BC085304.1 | 6 | 1199 | AAH85304.1 | |
| M28698.1 | 6 | 1194 | AAA60432.1 |

Genomic Map