Assay Details
Target Gene Details
Entrez Gene ID: | 217011 |
Gene Name: | notchless homolog 1 (Drosophila) |
Gene Aliases: |
AL022765, BC018399, Nle, l11Jus1, l11Jus4 |
Location: |
Chr.11:82900768-82908445 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nle1 | NM_145431.2 | 13 | 1580 | NP_663406.2 |
| AI852473.1 | 1 | 199 | ||
| AK141230.1 | 13 | 1555 | BAE24602.1 | |
| AK170853.1 | 13 | 1553 | BAE42074.1 | |
| BC018399.1 | 13 | 1571 | AAH18399.1 | |
| BX631936.1 | 1 | 192 |

Genomic Map