Assay Details
Target Gene Details
Entrez Gene ID: | 276952 |
Gene Name: | RAS-like, family 10, member B |
Gene Aliases: |
B230331P10Rik, Gm729, VTS58635 |
Location: |
Chr.11:83409142-83421039 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasl10b | NM_001013386.2 | 2 | 752 | NP_001013404.1 |
| AK045997.1 | 2 | 427 | ||
| AK158941.1 | 2 | 753 | BAE34735.1 | |
| BC116407.2 | 2 | 291 |

Genomic Map