Assay Details
Target Gene Details
Entrez Gene ID: | 74008 |
Gene Name: | arylsulfatase G |
Gene Aliases: |
6330406P08Rik, AI846872 |
Location: |
Chr.11:109450845-109573330 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsg |
Target Gene Details
Entrez Gene ID: | 100503496 |
Gene Name: | uncharacterized LOC100503496 |
Gene Aliases: |
- |
Location: |
Chr.11:109440078-109453359 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100503496 | NR_040680.1 | |||
| AK085963.1 | ||||
| BY742256.1 |
Target Gene Details
Entrez Gene ID: | 104681 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 6 |
Gene Aliases: |
AW743111, ESTM12, MCT 6, MCT 7, MCT6 |
Location: |
Chr.11:109450855-109473596 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a6 | NM_001029842.1 | 6 | 3371 | NP_001025013.1 |
| NM_134038.2 | 6 | 3314 | NP_598799.1 | |
| AK032026.1 | 6 | 3374 | BAC27657.1 |
More Information
Set Membership: |
|

Genomic Map