Assay Details
Target Gene Details
Entrez Gene ID: | 67477 |
Gene Name: | abhydrolase domain containing 15 |
Gene Aliases: |
1300007F04Rik |
Location: |
Chr.11:77515117-77520628 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd15 | NM_026185.4 | 1 | 464 | NP_080461.3 |
| AK004929.1 | 1 | 460 | BAC25102.1 | |
| AK018696.1 | 1 | 80 | BAB31349.1 | |
| AK042032.1 | 1 | 80 | BAC31138.1 | |
| AK046590.1 | 1 | 465 | BAE20655.1 | |
| AK153809.1 | 1 | 461 | ||
| FJ154119.1 | 1 | 382 |
Target Gene Details
Entrez Gene ID: | 216964 |
Gene Name: | transformation related protein 53 inducible protein 13 |
Gene Aliases: |
2410019G02Rik, Tp53i13 |
Location: |
Chr.11:77508099-77515921 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53i13 |
More Information
Set Membership: |
|

Genomic Map