Assay Details
Target Gene Details
Entrez Gene ID: | 67477 |
Gene Name: | abhydrolase domain containing 15 |
Gene Aliases: |
1300007F04Rik |
Location: |
Chr.11:77515117-77520628 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd15 | NM_026185.4 | 2 | 2851 | NP_080461.3 |
| AK046590.1 | 2 | 2852 | BAE20655.1 | |
| AK153809.1 | 2 | 2848 | ||
| CJ304668.1 | 1 | 81 |
More Information
Set Membership: |
|

Genomic Map