Assay Details
Target Gene Details
Entrez Gene ID: | 57316 |
Gene Name: | C1D nuclear receptor co-repressor |
Gene Aliases: |
1110036E10Rik, AI875855, SUN-CoR |
Location: |
Chr.11:17257618-17269176 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1d | NM_020558.3 | 5 | 1631 | NP_065583.2 |
| AK028702.1 | 5 | 1638 | BAC26075.1 | |
| AK035169.1 | 5 | 1632 | BAC28967.1 | |
| BC005436.1 | 5 | 1627 | AAH05436.1 |
More Information
Set Membership: |
|

Genomic Map