Assay Details
Target Gene Details
Entrez Gene ID: | 637515 |
Gene Name: | NLR family, pyrin domain containing 1B |
Gene Aliases: |
ENSMUSG00000070390, Nalp1b, OTTMUSG00000006087, OTTMUSG00000006089 |
Location: |
Chr.11:71153014-71230774 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp1b | NM_001040696.1 | 3 | 1737 | NP_001035786.1 |
| BC141354.1 | 2 | 1638 | ||
| DQ117591.1 | 3 | 1737 | AAZ40517.1 | |
| DQ117592.1 | 2 | 1610 | AAZ40518.1 | |
| DQ117593.1 | 2 | 1610 | AAZ40519.1 | |
| DQ153214.1 | 3 | 1737 | AAZ40529.1 |

Genomic Map