Assay Details
Target Gene Details
Entrez Gene ID: | 30805 |
Gene Name: | solute carrier family 22 (organic cation transporter), member 4 |
Gene Aliases: |
Octn1 |
Location: |
Chr.11:53983123-54028662 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a4 | NM_019687.3 | 1 | 84 | NP_062661.1 |
| AB016257.1 | 1 | 92 | BAA36626.1 | |
| AK143595.1 | 1 | 98 | ||
| BC010590.1 | 1 | 84 | AAH10590.1 |
More Information
Set Membership: |
|

Genomic Map