Assay Details
Target Gene Details
Entrez Gene ID: | 77097 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 |
Gene Aliases: |
3526402J09Rik, 5730590C14Rik |
Location: |
Chr.11:105589986-105929304 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tanc2 | NM_181071.3 | 7 | 1014 | NP_851416.2 |
| AK051973.1 | 7 | 1010 |

Genomic Map